Learn More
Med Vet International McKesson Inoculating Loop with Needle, 10 µL, 16-FLWN, 25/Bag
Catalog No. 503043275
Encompass_Preferred
Inoculating Loop with Needle McKesson 10 ?L ABS Integrated Handle Sterile Sterilized by gamma irradiation Disposable for collection and inoculation by streaking or puncturing methods Flexible feel Smooth construction facilitates uniform streaking of culture media Loop combinations feature integrated loop/needle design, eliminating the need for a separate needle
Catalog No. 50-304-3275 Supplier Med Vet International Supplier No. 1175311BG
Inoculating Loop with Needle McKesson 10 ?L ABS Integrated Handle Sterile Sterilized by gamma irradiation Disposable for collection and inoculation by streaking or puncturing methods Flexible feel Smooth construction facilitates uniform streaking of culture media Loop combinations feature integrated loop/needle design, eliminating the need for a separate needle