Learn More
Med Vet International McKesson Inoculating Loop, Dark Blue,10 ?L, 20-Centimeter Length, 16-10RL, 25/Bag
Catalog No. 503043223
Encompass_Preferred
Inoculating Loop McKesson 10 ?L Polystyrene Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Rigid feel media type Integrated handle Sterilized by gamma irradiation Dark blue There are 25 calibrated inoculating loops in a tamper-evident resealable bag
Catalog No. 50-304-3223 Supplier Med Vet International Supplier No. 1175310BG
Inoculating Loop McKesson 10 ?L Polystyrene Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Rigid feel media type Integrated handle Sterilized by gamma irradiation Dark blue There are 25 calibrated inoculating loops in a tamper-evident resealable bag