Learn More
Electron Microscopy Sciences Polystyrene White Needle
SDP
Supplier:  Electron Microscopy Sciences 6244810
Encompass_Preferred
Precision molded loop made from polystyrene eliminating the need for flaming metal loops and exposure to airborne pathogens. Made from virgin raw material which complies with USP class VI requirements. Cross contamination due to improper sterilization is eliminated. Surface treated for enhanced droplet adhesion. Supplied sterile in easy-to-open packs of 10 or 25. Color coded for product identification. Problem-free inoculation with smooth loop surface. Calibration certificate available.
Catalog No. 50-321-40