Learn More
Electron Microscopy Sciences Loop & Needle Holder, 8"
SDP
Catalog No. 5026712
Encompass_Preferred
Made from durable, heat resistant brass with a fiberglass insulating sleeve over the gripping area. The threaded collet type collar holds all wire gauge
Catalog No. 50-267-12 Supplier Electron Microscopy Sciences Supplier No. 6244008
Made from durable, heat resistant brass with a fiberglass insulating sleeve over the gripping area. The threaded collet type collar holds all wire gauge