Learn More
Grainger Loop Round 1/2 Dia White PK1565
Supplier:  Grainger 16U770
Encompass_Preferred
The Grainger 16U770 is a 120V, 60Hz, 1/3 HP, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor with a 1.15 Service Factor. It has a 48Y Frame with a 1" diameter shaft and is totally enclosed fan cooled (TEFC) for protection against dust and water. This electric motor can be used for various industrial applications.
Catalog No. 50-276-7305