Learn More
Microsolv Technology Corporation Loop Sample Peek Rheodyne100Ul
Catalog No. 5001452472
Encompass_Preferred
Sample Loop, Rheodyne, 100ul, For HPLC Injectors, PEEK. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52472 Supplier Microsolv Technology Corporation Supplier No. 47001100
Sample Loop, Rheodyne, 100ul, For HPLC Injectors, PEEK. MicroSolv Brand. 1/EA.