Learn More
Microsolv Technology Corporation Loop Sample Peek Valco 100Ul
Catalog No. 5001452479
Encompass_Preferred
Sample Loop, Valco, 100ul, For HPLC Injectors, PEEK. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52479 Supplier Microsolv Technology Corporation Supplier No. 47002100
Sample Loop, Valco, 100ul, For HPLC Injectors, PEEK. MicroSolv Brand. 1/EA.