Learn More
Microsolv Technology Corporation Loop Sample Peek Valco 2Ul
Catalog No. 5001452474
Encompass_Preferred
Sample Loop, Valco, 2ul, For HPLC Injectors, PEEK. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52474 Supplier Microsolv Technology Corporation Supplier No. 47002002
Sample Loop, Valco, 2ul, For HPLC Injectors, PEEK. MicroSolv Brand. 1/EA.