Learn More
Cramer Loop Stool
Catalog No. 03155056
Change view
Click to view available options
Back Style:
Open
Catalog No. Back Style
03-155-056 Open
1 options
Catalog No. 03-155-056 Supplier Cramer Supplier No. LPHU3012
Add to Cart
Add to Cart
Modern labs call for technical seating designed to move work forward with strength, style, and smooth mobility – Loop delivers all three.
Organic shape Ergonomically sculpted to cradle and support the body with natural comfort. Reliable mobility Strong, lightweight continuous loop steel frame stands up to damage-inflicting impacts. Simple cleaning, soft curves Zero edges plus a chemical-resistant polypropylene back keep disinfecting simple. Loop blends strength with modern design and lightweight mobility, delivering durability, style, and ease for hard-working lab environments. Modern design + custom colors Sleek tube frame and dynamic color palette add style to your space. Purposeful craftmanship + affordability Premium components in the simplest form at a lower price.
Features
  • Loop's continuous loop steel frame is strong, durable, and lightweight
  • Soft edges buffer damage-inflicting impact to the chair and surrounding environment
  • Minimal joint hygienic design for easy cleaning and disinfection
  • Organic chair shape receives and supports the user's body
  • Chemically-resistant polypropylene back for ultimate cleanability
  • Color palette complements any lab environment
Construction
  • 14-gauge powder-coated steel frame
  • i+™ Polyurethane seat
  • 25 in. durable nylon base
  • Polypropylene back
  • Field-replaceable seat cushion
  • Glass-filled nylon seat pan
Specifications
Product Type High Stool
With Lumbar Support No
Footrest Yes
With Tilt No
With Casters Yes
With Armrest No
With Backrest Yes
Color Black, Gray
Back Style Open
Material Powder-coated Steel
Seating Type Stool with Back