Learn More
Medchemexpress LLC Loperamide-d6 hydrochloride | 1189469-46-2 | 99.07% | 1 MG
SDP
Catalog No. 5000407577
Encompass_Preferred
Loperamide-d6 (hydrochloride) is a deuterium-labeled Loperamide hydrochloride. Loperamide hydrochloride itself is an opioid receptor agonist used for the treatment of diarrhea. This compound is intended for research use only and is not sold to patients.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07577 Supplier Medchemexpress LLC Supplier No. HYB0418AS1MG
Loperamide-d6 (hydrochloride) is a deuterium-labeled Loperamide hydrochloride. Loperamide hydrochloride itself is an opioid receptor agonist used for the treatment of diarrhea. This compound is intended for research use only and is not sold to patients.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.