Learn More
Medchemexpress LLC Loperamide hydrochloride | 34552-83-5 | 99.8% | 1 ML
SDP
Catalog No. 5000406543
Encompass_Preferred
Loperamide hydrochloride (R-18553 hydrochloride) is an opioid receptor agonist and a selective, competitive human intestinal carboxylesterases (hiCE) inhibitor. It is primarily recognized for its anti-diarrheal effects and is extensively used in research settings to study its impact on various biological pathways.
  • Induces hallmarks of ER stress and autophagy in GBM cells and MEFs.
  • Decreases LC3B and GABARAP lipidation levels in ATF4 KO cells compared to WT cells.
  • Used in cell autophagy assays.
  • Store at 4°C, sealed and away from moisture.
  • For solutions, store at -80°C for up to 6 months or -20°C for 1 month, sealed and away from moisture.
  • Shipped at room temperature for durations less than 2 weeks.
Catalog No. 50-004-06543 Supplier Medchemexpress LLC Supplier No. HYB0418A10MM/1ML
Loperamide hydrochloride (R-18553 hydrochloride) is an opioid receptor agonist and a selective, competitive human intestinal carboxylesterases (hiCE) inhibitor. It is primarily recognized for its anti-diarrheal effects and is extensively used in research settings to study its impact on various biological pathways.
  • Induces hallmarks of ER stress and autophagy in GBM cells and MEFs.
  • Decreases LC3B and GABARAP lipidation levels in ATF4 KO cells compared to WT cells.
  • Used in cell autophagy assays.
  • Store at 4°C, sealed and away from moisture.
  • For solutions, store at -80°C for up to 6 months or -20°C for 1 month, sealed and away from moisture.
  • Shipped at room temperature for durations less than 2 weeks.