Learn More
Medchemexpress LLC Loperamide hydrochloride | 34552-83-5 | 99.8% | 100 MG
SDP
Catalog No. 5000407041
Encompass_Preferred
Loperamide hydrochloride is an opioid receptor agonist and a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor. It has an anti-diarrheal effect and can also induce hallmarks of ER stress and autophagy in GBM cells and MEFs.
  • Opioid receptor agonist
  • Selective and competitive human intestinal carboxylesterases (hiCE) inhibitor
  • Anti-diarrheal effect
Catalog No. 50-004-07041 Supplier Medchemexpress LLC Supplier No. HYB0418A100MG
Loperamide hydrochloride is an opioid receptor agonist and a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor. It has an anti-diarrheal effect and can also induce hallmarks of ER stress and autophagy in GBM cells and MEFs.
  • Opioid receptor agonist
  • Selective and competitive human intestinal carboxylesterases (hiCE) inhibitor
  • Anti-diarrheal effect