Learn More
Medchemexpress LLC Loperamide (hydrochloride) | 34552-83-5 | 99.8% | 25 MG
SDP
Catalog No. 5000450567
Encompass_Preferred
Loperamide hydrochloride is an opioid receptor agonist and a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor. It exhibits an anti-diarrheal effect and is for research use only.
  • Opioid receptor agonist
  • Selective and competitive human intestinal carboxylesterases (hiCE) inhibitor
  • Exhibits anti-diarrheal effect
  • Induces hallmarks of ER stress and autophagy in GBM cells and MEFs (in vitro)
  • Decreases LC3B and GABARAP lipidation levels in ATF4 KO cells compared to WT cells (in vitro)
Catalog No. 50-004-50567 Supplier Medchemexpress LLC Supplier No. 25MGHYB0418AR
Loperamide hydrochloride is an opioid receptor agonist and a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor. It exhibits an anti-diarrheal effect and is for research use only.
  • Opioid receptor agonist
  • Selective and competitive human intestinal carboxylesterases (hiCE) inhibitor
  • Exhibits anti-diarrheal effect
  • Induces hallmarks of ER stress and autophagy in GBM cells and MEFs (in vitro)
  • Decreases LC3B and GABARAP lipidation levels in ATF4 KO cells compared to WT cells (in vitro)