Learn More
Medchemexpress LLC Loperamide hydrochloride | 34552-83-5 | 99.8% | 500 MG
SDP
Catalog No. 5000407891
Encompass_Preferred
Loperamide hydrochloride is an opioid receptor agonist and a selective human intestinal carboxylesterases (hiCE) inhibitor, known for its anti-diarrheal effect. This compound has a molecular formula of C29H34Cl2N2O2 and a molecular weight of 513.50. It is intended for research use only and has not been fully validated for medical applications.
  • Opioid receptor agonist
  • Selective and competitive human intestinal carboxylesterases inhibitor
  • Demonstrates anti-diarrheal properties
  • Induces ER stress and autophagy in certain cell lines (in vitro)
  • Decreases specific protein lipidation levels in ATF4 KO cells (in vitro)
  • Suitable for laboratory research purposes
Catalog No. 50-004-07891 Supplier Medchemexpress LLC Supplier No. HYB0418A500MG
Loperamide hydrochloride is an opioid receptor agonist and a selective human intestinal carboxylesterases (hiCE) inhibitor, known for its anti-diarrheal effect. This compound has a molecular formula of C29H34Cl2N2O2 and a molecular weight of 513.50. It is intended for research use only and has not been fully validated for medical applications.
  • Opioid receptor agonist
  • Selective and competitive human intestinal carboxylesterases inhibitor
  • Demonstrates anti-diarrheal properties
  • Induces ER stress and autophagy in certain cell lines (in vitro)
  • Decreases specific protein lipidation levels in ATF4 KO cells (in vitro)
  • Suitable for laboratory research purposes