Learn More
Medchemexpress LLC Loperamide hydrochloride (Standard) | 34552-83-5 | 99.9% | 100 MG
SDP
Catalog No. 5000407792
Encompass_Preferred
Loperamide hydrochloride (Standard) is the analytical standard of Loperamide (hydrochloride). It acts as an opioid receptor agonist and is recognized as a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor, exhibiting an anti-diarrheal effect. This product is intended for research and analytical applications.
  • Used as a reference standard for assay purposes.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Suitable for techniques such as HPLC, GC, and MS.
Catalog No. 50-004-07792 Supplier Medchemexpress LLC Supplier No. HYB0418AR100MG
Loperamide hydrochloride (Standard) is the analytical standard of Loperamide (hydrochloride). It acts as an opioid receptor agonist and is recognized as a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor, exhibiting an anti-diarrheal effect. This product is intended for research and analytical applications.
  • Used as a reference standard for assay purposes.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Suitable for techniques such as HPLC, GC, and MS.