Learn More
Cayman Chemical Loperamide (hydrochloride), Purity: ≥98%, CAS: 34552-83-5, 50 mg
SDP
Catalog No. 502972575
Encompass_Preferred
An opiate which potently and selectively activates μ opioid receptors (Ki = 0.16 nM) exclusively in the periphery; has antidiarrheal action; blocks voltage-sensitive sodium channels (IC50 = 270 nM) and high voltage-activated calcium channels (IC50 = 900 nM); comparable to morphine in blocking peripheral pain
Catalog No. 50-297-2575 Supplier Cayman Chemical Supplier No. 1487550 MG
An opiate which potently and selectively activates μ opioid receptors (Ki = 0.16 nM) exclusively in the periphery; has antidiarrheal action; blocks voltage-sensitive sodium channels (IC50 = 270 nM) and high voltage-activated calcium channels (IC50 = 900 nM); comparable to morphine in blocking peripheral pain