Learn More
Cayman Chemical Isovitexin, Purity: ≥98%, CAS: 38953-85-4, 10 mg
SDP
Catalog No. 502988409
Encompass_Preferred
A C-glycosylated flavone; scavenges DPPH radicals in a cell-free assay (IC50 = 370 µg/ml); cytotoxic to HepG2, MCF-7, and HCT116 cancer cells at 50 and 100 µg/ml; inhibits LPS-induced production of TNF-α and IL-6 and decreases iNOS and COX-2 levels in RAW 264.7 cells at 50 µg/ml; reduces cytotoxicity induced by Aβ (25-35) in PC12 cells at 200 µg/ml
Catalog No. 50-298-8409 Supplier Cayman Chemical Supplier No. 3121210 MG
A C-glycosylated flavone; scavenges DPPH radicals in a cell-free assay (IC50 = 370 µg/ml); cytotoxic to HepG2, MCF-7, and HCT116 cancer cells at 50 and 100 µg/ml; inhibits LPS-induced production of TNF-α and IL-6 and decreases iNOS and COX-2 levels in RAW 264.7 cells at 50 µg/ml; reduces cytotoxicity induced by Aβ (25-35) in PC12 cells at 200 µg/ml