Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decon Labs, Inc. LopHene II ST Single-Serve Detergent
BIOPHARMA

Catalog No. 18800103 Shop All Decon Labs Products
This item is not returnable. View return policy
Catalog No. 18-800-103 Supplier Decon Labs, Inc. Supplier No. 8813

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LopHene II ST Single-Serve Detergent is a sterile, concentrated, one-step germicidal detergent that cleans, disinfects and deodorizes

  • Germicidal, pseudomonacidal, tuberculocidal, fungicidal, and virucidal
  • Can be used in a rotational disinfection program with CiDecon ST
  • Tested, double-bagged, gamma-rradiated and tested for sterility
  • Lot Specific Document detailing QC, irradation and sterility testing is shipped wtih each case
  • Pre-measured in 2oz. bottles to be mixed with 2 gallons of water

Specifications

Product Type EPA Registered Disinfectant
Sterility Sterile
Appearance Clear to Light Yellow
For Use With (Application) Disinfecting surfaces
Quantity 24 Bottles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.