Learn More
Medchemexpress LLC HY-14588 100mg , Lopinavir CAS:192725-17-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14588100MG
Encompass_Preferred
HY-14588 100mg Lopinavir CAS: 192725-17-0 Lopinavir (ABT-378) is a highly potent, selective peptidomimetic inhibitor of the HIV-1 protease, with Kis of 1.3 to 3.6 pM for wild-type and mutant HIV protease. Lopinavir acts by arresting maturation of HIV-1 thereby blocking its infectivity. Lopinavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 14.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9360