Learn More
Apexbio Technology LLC Lopinavir 192725-17-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062159
Encompass_Preferred
Lopinavir is an inhibitor targeting the human immunodeficiency virus (HIV) protease enzyme implicated in viral polyprotein cleavage and maturation Structurally analogous to ritonavir lopinavir binds to HIV protease and suppresses catalytic activity consequently blocking HIV replication It demonstrates potent inhibitory actions against both wild-type and protease-inhibitor-resistant HIV variants Reported inhibition constants (Ki) for lopinavir range between 1 3 and 3 6 pM with antiviral efficacy represented by an EC50 below 0 06 M against protease mutants such as Val82 variants Due to its low susceptibility to inhibition by human serum proteins lopinavir is frequently utilized in antiviral screening assays drug-resistance research and investigations into HIV enzymology and resistance mechanisms
Catalog No. 50-000-62159 Supplier Apexbio Technology LLC Supplier No. A82045.1
Lopinavir is an inhibitor targeting the human immunodeficiency virus (HIV) protease enzyme implicated in viral polyprotein cleavage and maturation Structurally analogous to ritonavir lopinavir binds to HIV protease and suppresses catalytic activity consequently blocking HIV replication It demonstrates potent inhibitory actions against both wild-type and protease-inhibitor-resistant HIV variants Reported inhibition constants (Ki) for lopinavir range between 1 3 and 3 6 pM with antiviral efficacy represented by an EC50 below 0 06 M against protease mutants such as Val82 variants Due to its low susceptibility to inhibition by human serum proteins lopinavir is frequently utilized in antiviral screening assays drug-resistance research and investigations into HIV enzymology and resistance mechanisms