Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lopinavir, 98%, Thermo Scientific Chemicals

Catalog No. AC456750010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456750010 1 g
AC456750050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456750010 Supplier Thermo Scientific Chemicals Supplier No. 456750010
Add to Cart

Lopinavir, CAS # 192725-17-0, is an antiretroviral drug of the amphetamine class.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Lopinavir is a retroviral protease inhibitor that acts as an antiretroviral and can prevent the propagation of human immunodeficiency virus (HIV) and hepatitis viruses within cells

• Lopinavir is a dicarboxylic acid diamide and a substituted form of amphetamine

Applications

• Lopinavir may be used in the laboratory study of other retroviruses

• Lopinavir is suitable for laboratory research of drug delivery systems; some studies have shown that the activity of lopinavir can be enhanced by alternative delivery methods such as microencapsulation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192725-17-0
Molecular Formula C37H48N4O5
Molecular Weight (g/mol) 628.81
MDL Number MFCD22628840
InChI Key KJHKTHWMRKYKJE-SUGCFTRWSA-N
PubChem CID 133109001
IUPAC Name N-[(2R,4R,5R)-5-[[2-(2,6-dimethylphenoxy)acetyl]amino]-4-hydroxy-1,6-diphenylhexan-2-yl]-3-methyl-2-(2-oxo-1,3-diazinan-1-yl)butanamide
SMILES CC(C)[C@H](N1CCCNC1=O)C(=O)N[C@H](C[C@H](O)[C@H](CC1=CC=CC=C1)NC(=O)COC1=C(C)C=CC=C1C)CC1=CC=CC=C1

Specifications

Quantity 1 g
Chemical Name or Material Lopinavir

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.