Learn More
Selleck Chemical LLC Lopinavir 10mg 192725-17-0 ABT-378
SDP
Catalog No. 507876
Encompass_Preferred
Lopinavir (ABT-378) is a potent HIV protease inhibitor with Ki of 1.3 pM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-787-6 Supplier Selleck Chemical LLC Supplier No. S138010MG
Lopinavir (ABT-378) is a potent HIV protease inhibitor with Ki of 1.3 pM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.