Learn More
Aviva Systems Biology Corp LOR Antibody - N-terminal region (ARP41738_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41738_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LOR. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human LOR. Alternate names: MGC111513ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Mouse
Catalog No. 50-014-57973