Learn More
Medchemexpress LLC Loratadine-d5 | 1398065-63-8 | 99.96% | 387.91 | 5 MG
SDP
Catalog No. 5000385524
Encompass_Preferred
Loratadine-d5 is the deuterium labeled Loratadine. Loratadine (SCH-29851) is a selective inverse peripheral histamine H1-receptor agonist with an IC50 of >32 μM. It has anti-dengue-virus (DENV) activity and can inhibit immunologic release of inflammatory mediators. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Selective inverse peripheral histamine H1-receptor agonist.
  • Has anti-dengue-virus (DENV) activity.
  • Can inhibit immunologic release of inflammatory mediators.
  • Can be used as a tracer or an internal standard for quantitative analysis.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-85524 Supplier Medchemexpress LLC Supplier No. HY17043S15MG
Loratadine-d5 is the deuterium labeled Loratadine. Loratadine (SCH-29851) is a selective inverse peripheral histamine H1-receptor agonist with an IC50 of >32 μM. It has anti-dengue-virus (DENV) activity and can inhibit immunologic release of inflammatory mediators. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Selective inverse peripheral histamine H1-receptor agonist.
  • Has anti-dengue-virus (DENV) activity.
  • Can inhibit immunologic release of inflammatory mediators.
  • Can be used as a tracer or an internal standard for quantitative analysis.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.