Learn More
Cayman Chemical Loratadine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2748435-73-4, 1 mg
SDP
Catalog No. 502934949
Encompass_Preferred
An internal standard for the quantification of loratadine by GC- or LC-MS
Catalog No. 50-293-4949 Supplier Cayman Chemical Supplier No. 267591 MG
An internal standard for the quantification of loratadine by GC- or LC-MS