Learn More
Cayman Chemical LoratdIn-d4 500ug
SDP
Catalog No. 502936953
Encompass_Preferred
An internal standard for the quantification of loratadine by GC- or LC-MS
Catalog No. 50-293-6953 Supplier Cayman Chemical Supplier No. 26759500 UG
An internal standard for the quantification of loratadine by GC- or LC-MS