Learn More
Medchemexpress LLC HY-105272 10mg Medchemexpress, Loreclezole CAS:117857-45-1 Purity:>98%
SDP
Catalog No. 501968263
Encompass_Preferred
Medchemexpress, HY-105272 10mg Loreclezole CAS:117857-45-1 Loreclezole, an antiepileptic compound, is a selective GABAA receptor modulator and acts as a positive allosteric modulator of β2 or β3-subunit containing receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8263 Supplier Medchemexpress LLC Supplier No. HY10527210MG
Medchemexpress, HY-105272 10mg Loreclezole CAS:117857-45-1 Loreclezole, an antiepileptic compound, is a selective GABAA receptor modulator and acts as a positive allosteric modulator of β2 or β3-subunit containing receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.