Learn More
Enterprise Technology Solutions Lorell Desk Shell, Rectangular, 66"x30"x29-1/2", Mahogany
SDP
Catalog No. 5000791743
Encompass_Preferred
Rectangular desk shell is part of Lorell Essentials Series Laminate Furniture with high-quality laminate construction and metal-to-metal, cam-lock connections. Mahogany laminate top is 1" thick for durability. Use the two grommet holes in the top for easy cord routing. Design also includes a full modesty panel for privacy, 3mm PVC edges and adjustable feet.
Catalog No. 50-007-91743 Supplier Enterprise Technology Solutions Supplier No. LLR69372
Rectangular desk shell is part of Lorell Essentials Series Laminate Furniture with high-quality laminate construction and metal-to-metal, cam-lock connections. Mahogany laminate top is 1" thick for durability. Use the two grommet holes in the top for easy cord routing. Design also includes a full modesty panel for privacy, 3mm PVC edges and adjustable feet.