Learn More
Enterprise Technology Solutions Lorell Easel, w/Dry-Erase Board, Magnetic, 40"-70"H, Black
SDP
Catalog No. 5000791550
Encompass_Preferred
Magnetic Dry-erase Board Easel features dry-erase writing surface with an accessory tray. The adjustable height allows it to be used on a tabletop or as a floor-standing easel. Height adjusts from 40" to 70". Twist lock holds adjustable tubular legs in place. The 28" x 34" erasable surface offers a versatile way to present. Write, erase and write again. You can also post notes with magnets on the writing surface. Dry-erase board easel also offers anti-skid rubber feet for stability.
Catalog No. 50-007-91550 Supplier Enterprise Technology Solutions Supplier No. LLR75684
Magnetic Dry-erase Board Easel features dry-erase writing surface with an accessory tray. The adjustable height allows it to be used on a tabletop or as a floor-standing easel. Height adjusts from 40" to 70". Twist lock holds adjustable tubular legs in place. The 28" x 34" erasable surface offers a versatile way to present. Write, erase and write again. You can also post notes with magnets on the writing surface. Dry-erase board easel also offers anti-skid rubber feet for stability.