Learn More
Enterprise Technology Solutions Lorell Folding Table Cart, 75-7/8"x30-1/4"x45-1/4", Charcoal
SDP
Catalog No. 5000791370
Encompass_Preferred
Table cart stores and transports blow-molded rectangular tables up to 96" long. Extractable vertical posts around the trolley immobilize the stacked tables. Wheels have individual brakes for sure stopping while carrying heavy loads. The sturdy steel construction features a powder-coat finish for durability. Cart holds up to 20 tables with a 60" length; 20 tables with a 72" length; or 18 tables with a 96" length.
Catalog No. 50-007-91370 Supplier Enterprise Technology Solutions Supplier No. LLR65956
Table cart stores and transports blow-molded rectangular tables up to 96" long. Extractable vertical posts around the trolley immobilize the stacked tables. Wheels have individual brakes for sure stopping while carrying heavy loads. The sturdy steel construction features a powder-coat finish for durability. Cart holds up to 20 tables with a 60" length; 20 tables with a 72" length; or 18 tables with a 96" length.