Learn More
Enterprise Technology Solutions Lorell Monitor Arm, Dual, w/Pole, w/3.0 USB, Adjustable, Gray
SDP
Catalog No. 5000791279
Encompass_Preferred
Dual-monitor arm with a pole offers versatile positioning and useful features. Adjustable swivel and tilt enhance viewing flexibility. Arms swivel 135 degrees from side to side while the monitor mounting plate swivels 90 degrees. Face of the mounting plate also rotates 90 degrees. You can also tilt your monitor from negative 90 degrees to positive 90 degrees for multiple monitor viewing angles. Monitor arm is height-adjustable to easily improve eye level and posture for optimal comfort. Built-in spring gauge can be adjusted to suit various monitor weights. Two 3.0 USB ports on the pole are convenient for data transfer and charging. Rear tool holder allows easy access for any future adjustment. Detachable plates make monitor installation quick and easy. The option of top/side or grommet mounting makes installation convenient and quick. Built-in cable management and clip on pole help keep your cords organized.
Catalog No. 50-007-91279 Supplier Enterprise Technology Solutions Supplier No. LLR99803
Dual-monitor arm with a pole offers versatile positioning and useful features. Adjustable swivel and tilt enhance viewing flexibility. Arms swivel 135 degrees from side to side while the monitor mounting plate swivels 90 degrees. Face of the mounting plate also rotates 90 degrees. You can also tilt your monitor from negative 90 degrees to positive 90 degrees for multiple monitor viewing angles. Monitor arm is height-adjustable to easily improve eye level and posture for optimal comfort. Built-in spring gauge can be adjusted to suit various monitor weights. Two 3.0 USB ports on the pole are convenient for data transfer and charging. Rear tool holder allows easy access for any future adjustment. Detachable plates make monitor installation quick and easy. The option of top/side or grommet mounting makes installation convenient and quick. Built-in cable management and clip on pole help keep your cords organized.