Learn More
Enterprise Technology Solutions Lorell Pneumatic Height Stool, Backless, 24"x24"x23", Black
SDP
Catalog No. 5000791688
Encompass_Preferred
Backless stool features a 16" round, vinyl-covered seat, 2" dual-wheel casters, and a chrome five-star base. Functions include pneumatic seat-height adjustment from 17" to 22-1/2" and 360-degree swivel. Height adjusts easily in a 5" range with a lever under the seat. Stool holds up to 275 lb.
Catalog No. 50-007-91688 Supplier Enterprise Technology Solutions Supplier No. LLR69513
Backless stool features a 16" round, vinyl-covered seat, 2" dual-wheel casters, and a chrome five-star base. Functions include pneumatic seat-height adjustment from 17" to 22-1/2" and 360-degree swivel. Height adjusts easily in a 5" range with a lever under the seat. Stool holds up to 275 lb.