Learn More
Enterprise Technology Solutions Lorell Shelving,Wire,4 Shelves/4 Posts,36"x18"x72",Black
SDP
Catalog No. 5000791556
Encompass_Preferred
Industrial wire shelving is designed to provide the essentials for industrial wire shelving so you can organize your storage areas. Kit includes four shelves and four posts. Easy collar-and-sleeve assembly requires no tools. Each shelf adjusts in 1" increments. Shelving is made of steel with a chrome powder-coat finish, 10-gauge shelf surface, 3-gauge frame and 6-gauge wave pattern. Weight capacity is 1,000 lb. evenly distributed.
Catalog No. 50-007-91556 Supplier Enterprise Technology Solutions Supplier No. LLR70060
Industrial wire shelving is designed to provide the essentials for industrial wire shelving so you can organize your storage areas. Kit includes four shelves and four posts. Easy collar-and-sleeve assembly requires no tools. Each shelf adjusts in 1" increments. Shelving is made of steel with a chrome powder-coat finish, 10-gauge shelf surface, 3-gauge frame and 6-gauge wave pattern. Weight capacity is 1,000 lb. evenly distributed.