Learn More
Enterprise Technology Solutions Lorell Wall Clock, 13-1/4", Arabic Numerals, Black Frame
SDP
Catalog No. 5000791759
Encompass_Preferred
13-1/4" round wall clock features Quartz movement and a black frame. Easy-to-read white dial has Arabic numerals, black minute and hour hands, a red second hand, PVC clock face and glass front lens. Wall clock runs on one AA battery (sold separately).
Catalog No. 50-007-91759 Supplier Enterprise Technology Solutions Supplier No. LLR60989
13-1/4" round wall clock features Quartz movement and a black frame. Easy-to-read white dial has Arabic numerals, black minute and hour hands, a red second hand, PVC clock face and glass front lens. Wall clock runs on one AA battery (sold separately).