Learn More
Medchemexpress LLC Lorglumide sodium salt | 1021868-76-7 | 99.2% | 10 MM * 1 ML
SDP
Catalog No. 5000413686
Encompass_Preferred
Lorglumide sodium salt (CR-1409 sodium salt) is a potent cholecystokinin (CCK) receptor antagonist. It is intended for research use only.
  • Potent cholecystokinin (CCK) receptor antagonist.
  • For research use only.
Catalog No. 50-004-13686 Supplier Medchemexpress LLC Supplier No. HYB1439B10MM/1ML
Lorglumide sodium salt (CR-1409 sodium salt) is a potent cholecystokinin (CCK) receptor antagonist. It is intended for research use only.
  • Potent cholecystokinin (CCK) receptor antagonist.
  • For research use only.