Learn More
TARGETMOL CHEMICALS INC LORGLUMIDE SODIUM SALT 5MG
SDP
Catalog No. 502581507
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lorglumide sodium salt (CR-1409 sodium salt) is an antagonist of cholecystokinin receptors (CCK). purity: 99%
Catalog No. 50-258-1507 Supplier TARGETMOL CHEMICALS INC Supplier No. T118685MG
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lorglumide sodium salt (CR-1409 sodium salt) is an antagonist of cholecystokinin receptors (CCK). purity: 99%