Learn More
Cayman Chemical Lorglumide (sodium salt), Purity: ≥98%, CAS: 1021868-76-7, 10 mg
SDP
Catalog No. 502968176
Encompass_Preferred
A nonpeptidic antagonist of CCK1 (IC50 = 50 nM) that is 60-fold less effective at CCK2 (IC50 = 3 µM); blocks CCK-mediated gut muscle contraction, pancreatic growth, and pancreatic secretion; commonly used to study the roles of CCK1 in tissues and in animals
Catalog No. 50-296-8176 Supplier Cayman Chemical Supplier No. 1755510 MG
A nonpeptidic antagonist of CCK1 (IC50 = 50 nM) that is 60-fold less effective at CCK2 (IC50 = 3 µM); blocks CCK-mediated gut muscle contraction, pancreatic growth, and pancreatic secretion; commonly used to study the roles of CCK1 in tissues and in animals