Learn More
Medchemexpress LLC HY-12215 10mg Medchemexpress, Lorlatinib CAS:1454846-35-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1221510MG
Encompass_Preferred
Medchemexpress, HY-12215 10mg Lorlatinib CAS:1454846-35-5 Lorlatinib (PF-06463922) is a selective, orally active, brain-penetrant and ATP-competitive ROS1/ALK inhibitor. Lorlatinib has Kis of <0.025 nM, <0.07 nM, and 0.7 nM for ROS1, wild type ALK, and ALKL1196M, respectively. Lorlatinib has anticancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8862