Learn More
Selleck Chemical LLC Lorlatinib
SDP
Catalog No. 5000004099
Encompass_Preferred
Lorlatinib (PF-6463922) is a potent dual ALK/ROS1 inhibitor with Ki of 0 02 nM 0 07 nM and 0 7 nM for ROS1 ALK (WT) and ALK (L1196M) respectively PF-06463922 induces apoptosis Phase 1
Catalog No. 50-000-04099 Supplier Selleck Chemical LLC Supplier No. S753625MG
Lorlatinib (PF-6463922) is a potent dual ALK/ROS1 inhibitor with Ki of 0 02 nM 0 07 nM and 0 7 nM for ROS1 ALK (WT) and ALK (L1196M) respectively PF-06463922 induces apoptosis Phase 1