Learn More
TARGETMOL CHEMICALS INC LORLATINIB 25MG
SDP
Catalog No. 502580400
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lorlatinib (PF-6463922) is an orally available ATP-competitive inhibitor of the receptor tyrosine kinases anaplastic lymphoma kinase (ALK) and C-ros oncogene 1 (Ros1) with potential antineoplastic activity. purity: 99%
Catalog No. 50-258-0400 Supplier TARGETMOL CHEMICALS INC Supplier No. T306125MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lorlatinib (PF-6463922) is an orally available ATP-competitive inhibitor of the receptor tyrosine kinases anaplastic lymphoma kinase (ALK) and C-ros oncogene 1 (Ros1) with potential antineoplastic activity. purity: 99%