Learn More
Selleck Chemical LLC Lorlatinib(PF-6463922) 5mg 1454846-35-5
SDP
Catalog No. 501364913
Encompass_Preferred
Lorlatinib (PF-6463922) is a potent, dual ALK/ROS1 inhibitor with Ki of <0.02 nM, <0.07 nM, and 0.7 nM for ROS1, ALK (WT), and ALK (L1196M), respectively. PF-06463922 induces apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4913 Supplier Selleck Chemical LLC Supplier No. S75365MG
Add to Cart
Add to Cart
Lorlatinib (PF-6463922) is a potent, dual ALK/ROS1 inhibitor with Ki of <0.02 nM, <0.07 nM, and 0.7 nM for ROS1, ALK (WT), and ALK (L1196M), respectively. PF-06463922 induces apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.