Learn More
Sklar Instruments Clamp, Lorna-Edna Towel Clamp, Sklar 4 in., Non-Perforating, Curved, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501182770 Shop All Sklar Instruments Products
Encompass_Preferred
Clamp, Lorna-Edna Towel Clamp, Sklar, 4 in., Non-Perforating, Curved, Serrated, Stainless Steel, EA. Sklar Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, and surgical materials such as tubing and towels to the drape surrounding a surgical site. The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. The Lorna Towel Clamp is also commonly referred to as the Edna Towel Clamp. This clamp should never be used on skin. This product is 4 inches long. Lifetime warranty against all manufacturer defects.
Catalog No. 50-118-2770 Supplier Sklar Instruments Supplier No. 472837
Clamp, Lorna-Edna Towel Clamp, Sklar, 4 in., Non-Perforating, Curved, Serrated, Stainless Steel, EA. Sklar Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, and surgical materials such as tubing and towels to the drape surrounding a surgical site. The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. The Lorna Towel Clamp is also commonly referred to as the Edna Towel Clamp. This clamp should never be used on skin. This product is 4 inches long. Lifetime warranty against all manufacturer defects.