Learn More
Apexbio Technology LLC Lornoxicam 70374-39-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092515
Encompass_Preferred
Lornoxicam (CAS 70374-39-9) is an oxicam-class nonsteroidal anti-inflammatory drug (NSAID) that exerts its pharmacological effects primarily by potent inhibition of cyclooxygenase isoforms COX-1 and COX-2 with in vitro IC50 values of 5 nM and 8 nM respectively Studies in intact human cells and whole blood confirm its strong suppressive activity against both COX isoenzymes Additionally lornoxicam displays dose-dependent inhibition of nitric oxide formation (IC50 65 M) and reduction of IL-6 production in vitro Due to its robust COX inhibition and favorable pharmacokinetic profile lornoxicam is widely utilized as a research tool in studies investigating inflammation pain pathways and COX-mediated prostaglandin synthesis
Catalog No. 50-000-92515 Supplier Apexbio Technology LLC Supplier No. B14415.1
Lornoxicam (CAS 70374-39-9) is an oxicam-class nonsteroidal anti-inflammatory drug (NSAID) that exerts its pharmacological effects primarily by potent inhibition of cyclooxygenase isoforms COX-1 and COX-2 with in vitro IC50 values of 5 nM and 8 nM respectively Studies in intact human cells and whole blood confirm its strong suppressive activity against both COX isoenzymes Additionally lornoxicam displays dose-dependent inhibition of nitric oxide formation (IC50 65 M) and reduction of IL-6 production in vitro Due to its robust COX inhibition and favorable pharmacokinetic profile lornoxicam is widely utilized as a research tool in studies investigating inflammation pain pathways and COX-mediated prostaglandin synthesis