Learn More
Medchemexpress LLC Lornoxicam | 70374-39-9 | 98.7% | 1 ML
SDP
Catalog No. 5000406614
Encompass_Preferred
Lornoxicam is an orally active nonsteroidal anti-inflammatory drug (NSAID) that demonstrates analgesic, anti-inflammatory, antipyretic, and anticancer properties. It effectively inhibits both COX-1 and COX-2, as well as the production of NO by iNOS and the proinflammatory cytokine IL-6. This compound also inhibits tumor cell proliferation and migration and induces tumor cell apoptosis, making it suitable for research into inflammatory pain, colorectal cancer, and breast cancer.
  • Orally active nonsteroidal anti-inflammatory drug (NSAID)
  • Exhibits analgesic, anti-inflammatory, antipyretic, and anticancer activities
  • Effectively inhibits COX-1 and COX-2
  • Inhibits NO production by iNOS
  • Inhibits proinflammatory cytokine IL-6
  • Inhibits tumor cell proliferation and migration
  • Induces tumor cell apoptosis
  • Useful in inflammatory pain, colorectal cancer, and breast cancer studies
Catalog No. 50-004-06614 Supplier Medchemexpress LLC Supplier No. HYB036710MM/1ML
Lornoxicam is an orally active nonsteroidal anti-inflammatory drug (NSAID) that demonstrates analgesic, anti-inflammatory, antipyretic, and anticancer properties. It effectively inhibits both COX-1 and COX-2, as well as the production of NO by iNOS and the proinflammatory cytokine IL-6. This compound also inhibits tumor cell proliferation and migration and induces tumor cell apoptosis, making it suitable for research into inflammatory pain, colorectal cancer, and breast cancer.
  • Orally active nonsteroidal anti-inflammatory drug (NSAID)
  • Exhibits analgesic, anti-inflammatory, antipyretic, and anticancer activities
  • Effectively inhibits COX-1 and COX-2
  • Inhibits NO production by iNOS
  • Inhibits proinflammatory cytokine IL-6
  • Inhibits tumor cell proliferation and migration
  • Induces tumor cell apoptosis
  • Useful in inflammatory pain, colorectal cancer, and breast cancer studies