Learn More
Selleck Chemical LLC Losartan 25mg 114798-26-4
SDP
Supplier:  Selleck Chemical LLC S506725MG
Encompass_Preferred
Losartan is a selective, orally administered, nonpeptide blocker of angiotensin II type 1 (AT1) receptor used to treat high blood pressure, diabetic kidney disease, heart failure, and left ventricular enlargement. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8319