Learn More
Medchemexpress LLC HY-17512 500mg , Losartan CAS:114798-26-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17512500MG
Encompass_Preferred
HY-17512 500mg Losartan CAS: 114798-26-4 Losartan is an angiotensin II receptor antagonist, competing with the binding of angiotensin II to AT1 receptors with IC50 of 20 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9776