Learn More
Selleck Chemical LLC Losartan S5067-1g
SDP
Catalog No. 503112234
Encompass_Preferred
Losartan(DuP-753) is a selective orally administered nonpeptide blocker of angiotensin II type 1 (AT1) receptor used to treat high blood pressure diabetic kidney disease heart failure and left ventricular enlargement
Catalog No. 50-311-2234 Supplier Selleck Chemical LLC Supplier No. S50671G
Losartan(DuP-753) is a selective orally administered nonpeptide blocker of angiotensin II type 1 (AT1) receptor used to treat high blood pressure diabetic kidney disease heart failure and left ventricular enlargement