Learn More
Medchemexpress LLC Losartan Carboxylic Acid | 124750-92-1 | 99.5% | 10 MG
SDP
Catalog No. 5000340868
Encompass_Preferred
Losartan Carboxylic Acid (E-3174) is an active carboxylic acid metabolite of Losartan, functioning as an angiotensin II receptor type 1 (AT1) antagonist. It blocks angiotensin II-induced responses in vascular smooth muscle cells and elevates plasma renin activities while reducing mean arterial pressure.
  • Blocks angiotensin II-induced responses in vascular smooth muscle cells.
  • Elevates plasma renin activities.
  • Reduces mean arterial pressure.
  • Inhibits specific binding of angiotensin II to vascular smooth muscle cells with an IC50 of 1.1 nM.
  • Abolishes angiotensin II-induced formation of inositolphosphates in vascular smooth muscle cells.
  • Inhibits angiotensin II-induced elevation of intracellular cytosolic Ca2+ concentration with an IC50 of 5 nM.
  • Inhibits angiotensin II-induced cell protein synthesis with an IC50 of 3 nM.
  • Induces an 87±4% inhibition of pressor responses to angiotensin I in mongrel dogs at 0.1 mg/kg.
Catalog No. 50-003-40868 Supplier Medchemexpress LLC Supplier No. HY1276510MG
Losartan Carboxylic Acid (E-3174) is an active carboxylic acid metabolite of Losartan, functioning as an angiotensin II receptor type 1 (AT1) antagonist. It blocks angiotensin II-induced responses in vascular smooth muscle cells and elevates plasma renin activities while reducing mean arterial pressure.
  • Blocks angiotensin II-induced responses in vascular smooth muscle cells.
  • Elevates plasma renin activities.
  • Reduces mean arterial pressure.
  • Inhibits specific binding of angiotensin II to vascular smooth muscle cells with an IC50 of 1.1 nM.
  • Abolishes angiotensin II-induced formation of inositolphosphates in vascular smooth muscle cells.
  • Inhibits angiotensin II-induced elevation of intracellular cytosolic Ca2+ concentration with an IC50 of 5 nM.
  • Inhibits angiotensin II-induced cell protein synthesis with an IC50 of 3 nM.
  • Induces an 87±4% inhibition of pressor responses to angiotensin I in mongrel dogs at 0.1 mg/kg.