Learn More
Medchemexpress LLC Losartan carboxylic acid | 124750-92-1 | MFCD00871928 | 99.5% | 436.89 | C22H21ClN6O2 | 5 MG
SDP
Catalog No. 5000264457
Encompass_Preferred
Losartan carboxylic acid (E-3174) is the active carboxylic acid metabolite of losartan and a non-peptide angiotensin II type 1 (AT1) receptor antagonist. Supplied as a solid, high-purity reference standard, it is intended for research use in pharmacology, metabolite identification, and analytical method development.
  • High-purity solid reference suitable for analytical assays and QC.
  • Active AT1 receptor antagonist metabolite used in pharmacology studies.
  • Useful for metabolite identification and compound characterization.
  • Available in small milligram pack sizes for research applications.
  • Molecular formula C22H21ClN6O2 and molecular weight 436.89 g/mol.
Catalog No. 50-002-64457 Supplier Medchemexpress LLC Supplier No. HY127655MG
Losartan carboxylic acid (E-3174) is the active carboxylic acid metabolite of losartan and a non-peptide angiotensin II type 1 (AT1) receptor antagonist. Supplied as a solid, high-purity reference standard, it is intended for research use in pharmacology, metabolite identification, and analytical method development.
  • High-purity solid reference suitable for analytical assays and QC.
  • Active AT1 receptor antagonist metabolite used in pharmacology studies.
  • Useful for metabolite identification and compound characterization.
  • Available in small milligram pack sizes for research applications.
  • Molecular formula C22H21ClN6O2 and molecular weight 436.89 g/mol.