Learn More
TARGETMOL CHEMICALS INC LOSARTAN CARBOXYLIC ACID 25MG
SDP
Catalog No. 502580254
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Losartan Carboxylic Acid (E-3174) a potent AT1 antagonist (Kis 0.57 nM rat; 0.67 nM human) producing a depressor response and vasodilatation. Losartan Carboxylic Acid a metabolite of losartan is more potent than losartan in blocking the angiotensin II-induced responses in vascular smooth muscle cells. purity: 99%
Catalog No. 50-258-0254 Supplier TARGETMOL CHEMICALS INC Supplier No. T346125MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Losartan Carboxylic Acid (E-3174) a potent AT1 antagonist (Kis 0.57 nM rat; 0.67 nM human) producing a depressor response and vasodilatation. Losartan Carboxylic Acid a metabolite of losartan is more potent than losartan in blocking the angiotensin II-induced responses in vascular smooth muscle cells. purity: 99%